Learn More
Medchemexpress LLC Trazodone hydrochloride | 25332-39-2 | 99.9% | 25 MG
SDP
Catalog No. 5000450545
Encompass_Preferred
Trazodone hydrochloride (Standard) is an analytical standard and serotonin receptor antagonist and reuptake inhibitor (SARI). It is intended for research and analytical applications, and exhibits antidepressant and anti-insomnious activities. It also acts as an antagonist against α1- and α2-adrenergic receptors and histamine H1 receptors.
  • Analytical standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Serotonin receptor antagonist and reuptake inhibitor (SARI)
  • Exhibits antidepressant and anti-insomnious activities
  • Acts as an antagonist against α1- and α2-adrenergic receptors and histamine H1 receptors
  • Minimal anticholinergic effects
Catalog No. 50-004-50545 Supplier Medchemexpress LLC Supplier No. 25MGHYB0478R
Trazodone hydrochloride (Standard) is an analytical standard and serotonin receptor antagonist and reuptake inhibitor (SARI). It is intended for research and analytical applications, and exhibits antidepressant and anti-insomnious activities. It also acts as an antagonist against α1- and α2-adrenergic receptors and histamine H1 receptors.
  • Analytical standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Serotonin receptor antagonist and reuptake inhibitor (SARI)
  • Exhibits antidepressant and anti-insomnious activities
  • Acts as an antagonist against α1- and α2-adrenergic receptors and histamine H1 receptors
  • Minimal anticholinergic effects