Learn More
Medchemexpress LLC Trazodone hydrochloride | 25332-39-2 | MFCD00079603 | 99.6% | 1 G
SDP
Catalog No. 5000406384
Encompass_Preferred
Trazodone hydrochloride is an antidepressant belonging to the class of serotonin receptor antagonists and reuptake inhibitors (SARIs). It is a triazolopyridine derivative with antidepressant and anti-insomnious activity. Trazodone hydrochloride is often used for the treatment of anxiety disorders and depression. It exerts antagonistic properties against α1- and α2-adrenergic receptors.
  • Antidepressant and anti-insomnious activity.
  • Serotonin receptor antagonist and reuptake inhibitor.
  • Antagonizes α1- and α2-adrenergic receptors.
  • Used in the treatment of anxiety disorders and depression.
Catalog No. 50-004-06384 Supplier Medchemexpress LLC Supplier No. HYB04781G
Trazodone hydrochloride is an antidepressant belonging to the class of serotonin receptor antagonists and reuptake inhibitors (SARIs). It is a triazolopyridine derivative with antidepressant and anti-insomnious activity. Trazodone hydrochloride is often used for the treatment of anxiety disorders and depression. It exerts antagonistic properties against α1- and α2-adrenergic receptors.
  • Antidepressant and anti-insomnious activity.
  • Serotonin receptor antagonist and reuptake inhibitor.
  • Antagonizes α1- and α2-adrenergic receptors.
  • Used in the treatment of anxiety disorders and depression.