Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trazodone Hydrochloride, MP Biomedicals™

Catalog No. ICN19151801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19151801 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19151801 Supplier MP Biomedicals, Inc Supplier No. 0219151801

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25332-39-2
Molecular Formula C19H23Cl2N5O
Molecular Weight (g/mol) 408.327
InChI Key OHHDIOKRWWOXMT-UHFFFAOYSA-N
Synonym trazodone hydrochloride, trazodone hcl, desyrel, bimaran, molipaxin, devidon, pragmazone, thombran, tombran, tritico
PubChem CID 62935
ChEBI CHEBI:9655
IUPAC Name 2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride
SMILES C1CN(CCN1CCCN2C(=O)N3C=CC=CC3=N2)C4=CC(=CC=C4)Cl.Cl

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.