Learn More
CAPRICO BIOTECHNOLOGIES INC TRBC1 PE Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4133025
Encompass_Preferred
The TRBC1 Polyclonal Antibody from Caprico Biotechnologies is a rabbit polyclonal antibody raised against amino acids 1-100 mapping at the N-terminus of TRBC1 of human origin. This antibody is recommended for detection of TRBC1 of mouse, rat and human origin by Western Blot, Immunoprecipitation, and ELISA. The immunogen is a recombinant protein of human TRBC1. This antibody is purified by protein A and peptide affinity chromatography. The molecular weight of TRBC1 is approximately 55 kDa.
Catalog No. 50-227-9312