Learn More
CAPRICO BIOTECHNOLOGIES INC TRBC1 PE-Jovi 1 RUO
Supplier:  CAPRICO BIOTECHNOLOGIES INC 113325
Encompass_Preferred
The TRBC1 PE-Jovi 1 RUO is a recombinant polyclonal antibody produced in rabbits against the human TRBC1 protein. It is intended for research use only to detect TRBC1 protein via Western blot, immunohistochemistry, and immunofluorescence. This antibody is purified via protein A affinity chromatography and is supplied lyophilized from a 0.2 μm filtered solution in PBS. Each vial contains 100 μg of antibody.
Catalog No. NC2093539