Learn More
Cayman Chemical Trbulus trstrs L Gtogn 10mg
SDP
Catalog No. 502910860
Encompass_Preferred
A sapogenin with enzyme inhibitory and hormone-releasing activities; an inhibitor of UGT1A4 (IC50 = 0.69 µM); stimulates the release of growth hormone in primary rat pituitary cells at 20 UG/ml
Catalog No. 50-291-0860 Supplier Cayman Chemical Supplier No. 3468110 MG
A sapogenin with enzyme inhibitory and hormone-releasing activities; an inhibitor of UGT1A4 (IC50 = 0.69 µM); stimulates the release of growth hormone in primary rat pituitary cells at 20 UG/ml