Learn More
Cayman Chemical Gitogenin, Purity: ≥98%, CAS: 511-96-6, 25 mg
SDP
Catalog No. 502911359
Encompass_Preferred
A sapogenin with enzyme inhibitory and hormone-releasing activities; an inhibitor of UGT1A4 (IC50 = 0.69 µM); stimulates the release of growth hormone in primary rat pituitary cells at 20 µg/ml
Catalog No. 50-291-1359 Supplier Cayman Chemical Supplier No. 3468125 MG
A sapogenin with enzyme inhibitory and hormone-releasing activities; an inhibitor of UGT1A4 (IC50 = 0.69 µM); stimulates the release of growth hormone in primary rat pituitary cells at 20 µg/ml