Learn More
SPEX CERTIPREP LLC 3P Total Residual Chlorine Concentrate CRM 1.50 mg/L, 24/pk
Catalog No. NC0457934
Encompass_Preferred
(NSI by Zeptometrix) Convenient and easy to use. Dilute 1mL from an ampule to 1000mL to reach certified value. Always the same, lot to lot, so your dilution process remains the same minimizing errors in your lab.
Catalog No. NC0457934 Supplier SPEX CERTIPREP LLC 3P Supplier No. QCI188
(NSI by Zeptometrix) Convenient and easy to use. Dilute 1mL from an ampule to 1000mL to reach certified value. Always the same, lot to lot, so your dilution process remains the same minimizing errors in your lab.