Learn More
SPEX CERTIPREP LLC 3P TRC CONCENTRATE 1MG/L 24/PK
Catalog No. NC0457933
Encompass_Preferred
(NSI by Zeptometrix) Total Residual Chlorine Concentrate CRM, 24 ampules, 1.00 mg/L, Dilutes to 1:1000
Catalog No. NC0457933 Supplier SPEX CERTIPREP LLC 3P Supplier No. QCI187
(NSI by Zeptometrix) Total Residual Chlorine Concentrate CRM, 24 ampules, 1.00 mg/L, Dilutes to 1:1000