Learn More
SPEX CERTIPREP LLC 3P TRC CONCENTRATE .5MG/L 24/PK
Catalog No. NC0457932
Encompass_Preferred
(NSI by Zeptometrix) Total Residual Chlorine Concentrate CRM, 24 ampules, 0.500mg/L, Dilutes to 1:1000
Catalog No. NC0457932 Supplier SPEX CERTIPREP LLC 3P Supplier No. QCI184
(NSI by Zeptometrix) Total Residual Chlorine Concentrate CRM, 24 ampules, 0.500mg/L, Dilutes to 1:1000