Learn More
Selleck Chemical LLC TRC102-E1503-250MG
SDP
Catalog No. 503138241
Encompass_Preferred
TRC102 (methoxyamine hydrochloride O-Methylhydroxylamine hydrochloride) is a reagent used for the preparation of O-methyl oximes or the synthesis of O-methyl oximes from aldehydes
Catalog No. 50-313-8241 Supplier Selleck Chemical LLC Supplier No. E1503250MG
TRC102 (methoxyamine hydrochloride O-Methylhydroxylamine hydrochloride) is a reagent used for the preparation of O-methyl oximes or the synthesis of O-methyl oximes from aldehydes