Learn More
Cayman Chemical Torcetrapib, Purity: ≥98%, CAS: 262352-17-0, 50 mg
SDP
Catalog No. 502970208
Encompass_Preferred
A CETP inhibitor (IC50s = 47 and 51 nM for cholesteryl and triglyceride transfer, respectively); selective for CETP over PLTP, LCAT, and hepatic and lipoprotein lipases at 10 µM; inhibits CETP activity and increases HDL cholesterol levels in serum from transgenic mice expressing human CETP and ApoA1 at 30 mg/kg; increases mean arterial blood pressure in Zucker fa/fa diabetic rats at 60 and 200 mg/kg
Catalog No. 50-297-0208 Supplier Cayman Chemical Supplier No. 1502850 MG
A CETP inhibitor (IC50s = 47 and 51 nM for cholesteryl and triglyceride transfer, respectively); selective for CETP over PLTP, LCAT, and hepatic and lipoprotein lipases at 10 µM; inhibits CETP activity and increases HDL cholesterol levels in serum from transgenic mice expressing human CETP and ApoA1 at 30 mg/kg; increases mean arterial blood pressure in Zucker fa/fa diabetic rats at 60 and 200 mg/kg