Learn More
Invivogen Plasmocin - Mycoplasma Elimination Reagent, 50 mg (2 x 1 ml)
SDP
Catalog No. NC9698402
Encompass
2ml liquid at 25mg/ml packaging is 2 - 1ml tubes for total of 50mg. for treatment
Catalog No. NC9698402 Supplier Invivogen Supplier No. ANTMPT
2ml liquid at 25mg/ml packaging is 2 - 1ml tubes for total of 50mg. for treatment