Learn More
Biosurface Technologies Corp Treatment Imaging Flow Cell, for Imaging of Biofilms on Disc Coupons During Treatment.
Catalog No. NC3662432
Encompass
"The Model FC 310 Treatment Imaging Flow Cell was designed specifically for the Center for Biofilm Engineering to provide imaging from the top of the biofilm during treatment with biocides, removal agents, and other chemical agents. This unique perspective provides imaging of the irregular biofilm surface as it comes into contact with the treatment chemistry. The small liquid volume of the cell minimizes the use of valuable chemical compounds. While this flow cell was designed to provide quick installation and access of the disc coupons, allowing installation of disc coupons with attached biofilm (biofilm grown in the CDC, RDR, or Bio-inLine Biofilm Reactors), it can also be used to image developing biofilms.The FC 310 Treatment Imaging Flow Cell is machined from polycarbonate plastic and is completely autoclavable and reusable.
Catalog No. NC3662432 Supplier Biosurface Technologies Corp Supplier No. FC 310
"The Model FC 310 Treatment Imaging Flow Cell was designed specifically for the Center for Biofilm Engineering to provide imaging from the top of the biofilm during treatment with biocides, removal agents, and other chemical agents. This unique perspective provides imaging of the irregular biofilm surface as it comes into contact with the treatment chemistry. The small liquid volume of the cell minimizes the use of valuable chemical compounds. While this flow cell was designed to provide quick installation and access of the disc coupons, allowing installation of disc coupons with attached biofilm (biofilm grown in the CDC, RDR, or Bio-inLine Biofilm Reactors), it can also be used to image developing biofilms.The FC 310 Treatment Imaging Flow Cell is machined from polycarbonate plastic and is completely autoclavable and reusable.