Learn More
Trebuchet Kits
SCI_ED
Catalog No. S94039
Change view
Click to view available options
Single
Catalog No.
S94039 Single
1 options
This item is not returnable. View return policy
Catalog No. S94039 Supplier Envision System Inc Supplier No. 10200
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Instructions and internet resource materials for investigations
Fun and functional.
Useful in studying energy conversion, levers, trajectories, ratios, rotational motion, inertia, forces and more. Investigate how projectile range is altered by changing variables such as weight, weight position, sling length, pivot point and projectile mass.
  • Powered by gravitational potential energy
  • Easy to assemble
  • Provides hands-on building experience
Specifications
Product Type Kit, Trebuchet
For Use With (Application) Studying energy conversion, levers, trajectories, ratios, rotational motion, inertia, forces and more
Includes 1 Precut Wooden sheet, 6 Metal washers, 1 Metal rod 3.75 in. L, 1 wire 5 in. L, 1 Cloth rectangle, 1 packet of clay, 1 Rubber band, 36 in. monofilamen