Learn More
LABSYSTEMS DIAGNOSTICS OY Multilevel Control in DBS format
Catalog No. NC2170392
Encompass_Preferred
One DBS card, each with four control spots, packed in resealable foil pouche with desiccant. The DBS card is Whatman #903 filter paper, on which whole blood spiked with different levels of SMN1, TREC and RNase P plasmid DNA is spotted. The TREC-SMN1-RNase P DBS Control can be used with SCID-SMA Multiplex Real Time PCR assay
This item is not returnable. View return policy
Catalog No. NC2170392 Supplier LABSYSTEMS DIAGNOSTICS OY Supplier No. 34699309
One DBS card, each with four control spots, packed in resealable foil pouche with desiccant. The DBS card is Whatman #903 filter paper, on which whole blood spiked with different levels of SMN1, TREC and RNase P plasmid DNA is spotted. The TREC-SMN1-RNase P DBS Control can be used with SCID-SMA Multiplex Real Time PCR assay