Learn More
Medchemexpress LLC Trecovirsen (sodium) | 170274-79-0 | >=98% | 7776.30 | 1 MG
SDP
Catalog No. 5000360065
Encompass_Preferred
Trecovirsen is an antiviral agent that targets HIV gag mRNA by hybridizing with complementary HIV gag mRNA at the initiation site. It induces a reversible, dose-dependent prolongation of activated partial thromboplastin time due to its polyanionic properties and is applicable for research related to HIV infection.
  • Induces a reversible, dose-dependent prolongation of activated partial thromboplastin time.
  • Applicable for research related to HIV infection.
  • Produces potent, long-lasting inhibitory effects against HIV in cultured HIV-infected cells.
  • Potently inhibits HIV-1 replication in various in vitro cell models.
  • Exhibits widespread tissue distribution, with highest accumulation in the liver and kidney.
Catalog No. 50-003-60065 Supplier Medchemexpress LLC Supplier No. HY149906C1MG
Trecovirsen is an antiviral agent that targets HIV gag mRNA by hybridizing with complementary HIV gag mRNA at the initiation site. It induces a reversible, dose-dependent prolongation of activated partial thromboplastin time due to its polyanionic properties and is applicable for research related to HIV infection.
  • Induces a reversible, dose-dependent prolongation of activated partial thromboplastin time.
  • Applicable for research related to HIV infection.
  • Produces potent, long-lasting inhibitory effects against HIV in cultured HIV-infected cells.
  • Potently inhibits HIV-1 replication in various in vitro cell models.
  • Exhibits widespread tissue distribution, with highest accumulation in the liver and kidney.