Learn More
Waters Corp Trefoil AMY1 Column, 2.5 μm, 2.1 mm ID, 150 mm
Catalog No. 502392113 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Trefoil 2.5 μm Columns are optimized for ACQUITY UPC2 systems for chiral separations. They enable faster, more efficient analysis with mass spectrometer-compatible small particles. Complementary chemistries offer distinct retention and improved selectivity for distinguishing chiral molecules, separating enantiomers, stereoisomers, metabolites, degradants, and contaminants with enhanced precision and speed.
  • Optimized for ACQUITY UPC2 system.
  • Enables faster, more efficient chiral analysis.
  • Mass spectrometer compatible.
  • Distinct retention for chiral molecules.
  • Enhanced selectivity via modifiers.
  • Separates enantiomers, stereoisomers, metabolites, degradants, contaminants precisely.
Catalog No. 50-239-2113 Supplier Waters Corp Supplier No. 186007458
Trefoil 2.5 μm Columns are optimized for ACQUITY UPC2 systems for chiral separations. They enable faster, more efficient analysis with mass spectrometer-compatible small particles. Complementary chemistries offer distinct retention and improved selectivity for distinguishing chiral molecules, separating enantiomers, stereoisomers, metabolites, degradants, and contaminants with enhanced precision and speed.
  • Optimized for ACQUITY UPC2 system.
  • Enables faster, more efficient chiral analysis.
  • Mass spectrometer compatible.
  • Distinct retention for chiral molecules.
  • Enhanced selectivity via modifiers.
  • Separates enantiomers, stereoisomers, metabolites, degradants, contaminants precisely.