Learn More
Waters Corp Column, AMY1, 2.5 μm, 3 mm X 50 mm
Catalog No. 502392278 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 2.5 μm columns for chiral separations are designed to reduce method development time while providing optimum peak shape and efficiencies. They are optimized for the ACQUITY UPC2 System, enabling faster and more efficient chiral method development and analysis with the added performance of small particles and compatibility with a full range of mass spectrometers.
  • Designed for chiral separations.
  • Reduces method development time.
  • Provides optimum peak shape and efficiencies.
  • Optimized for ACQUITY UPC2 System.
  • Enables faster and more efficient chiral method development and analysis.
  • Features small particles for added performance.
  • Compatible with a full range of mass spectrometers.
Catalog No. 50-239-2278 Supplier Waters Corp Supplier No. 186007459
These 2.5 μm columns for chiral separations are designed to reduce method development time while providing optimum peak shape and efficiencies. They are optimized for the ACQUITY UPC2 System, enabling faster and more efficient chiral method development and analysis with the added performance of small particles and compatibility with a full range of mass spectrometers.
  • Designed for chiral separations.
  • Reduces method development time.
  • Provides optimum peak shape and efficiencies.
  • Optimized for ACQUITY UPC2 System.
  • Enables faster and more efficient chiral method development and analysis.
  • Features small particles for added performance.
  • Compatible with a full range of mass spectrometers.