Learn More
Waters Corp Column, AMY1, 2.5 μm, 3 mm x 150 mm
Catalog No. 502391014 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Trefoil 2.5 μm Columns for chiral separations are designed to reduce method development time while providing optimum peak shape and efficiencies. Optimized for ACQUITY UPC2 System, these columns allow for faster and more efficient chiral method development and analysis with the added performance of small particles and compatibility with a full range of mass spectrometers.
  • Designed to reduce method development time while providing optimum peak shape and efficiencies for chiral separations.
  • Optimized for ACQUITY UPC2 System.
  • Achieve faster and more efficient chiral method development and analysis.
  • Added performance of columns packed with small particles.
  • Compatible with a full range of mass spectrometers.
Catalog No. 50-239-1014 Supplier Waters Corp Supplier No. 186007460
Trefoil 2.5 μm Columns for chiral separations are designed to reduce method development time while providing optimum peak shape and efficiencies. Optimized for ACQUITY UPC2 System, these columns allow for faster and more efficient chiral method development and analysis with the added performance of small particles and compatibility with a full range of mass spectrometers.
  • Designed to reduce method development time while providing optimum peak shape and efficiencies for chiral separations.
  • Optimized for ACQUITY UPC2 System.
  • Achieve faster and more efficient chiral method development and analysis.
  • Added performance of columns packed with small particles.
  • Compatible with a full range of mass spectrometers.