Learn More
Waters Corp Column, CEL1, 2.5 μm, 2.1 mm x 150 mm, 1 pk
Catalog No. 502391974 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are designed for chiral separations, optimized for SuperCritical Fluid Chromatography (SFC) systems. They provide optimum peak shape and efficiencies, enabling faster, more efficient method development and analysis. Manufactured under controlled conditions, these modified polysaccharide-based stationary phases offer broad-spectrum chiral selectivity.
  • Reduces method development time
  • Provides optimum peak shape and efficiencies
  • Compatible with a full range of mass spectrometers
  • Manufactured under stringent quality assurance standards
  • Offers broad-spectrum chiral selectivity
  • Ensures high quality, consistent, and reproducible results
  • Separates enantiomers, stereoisomers, metabolites, degradants, and impurities
  • Supports cost-effective methods using compressed liquid CO2
Catalog No. 50-239-1974 Supplier Waters Corp Supplier No. 186007462
These columns are designed for chiral separations, optimized for SuperCritical Fluid Chromatography (SFC) systems. They provide optimum peak shape and efficiencies, enabling faster, more efficient method development and analysis. Manufactured under controlled conditions, these modified polysaccharide-based stationary phases offer broad-spectrum chiral selectivity.
  • Reduces method development time
  • Provides optimum peak shape and efficiencies
  • Compatible with a full range of mass spectrometers
  • Manufactured under stringent quality assurance standards
  • Offers broad-spectrum chiral selectivity
  • Ensures high quality, consistent, and reproducible results
  • Separates enantiomers, stereoisomers, metabolites, degradants, and impurities
  • Supports cost-effective methods using compressed liquid CO2