Learn More
Waters Corp Column, Trefoil CEL2, 2.5 μm, 2.1 mm X 50 mm
Catalog No. 502390768 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These 2.5 μm columns for chiral separations are designed to reduce method development time while providing optimum peak shape and efficiencies. They are optimized for ACQUITY UPC2 Systems, allowing for faster and more efficient chiral method development and analysis with the added performance of columns packed with small particles and compatibility with a full range of mass spectrometers.
  • Designed to reduce method development time
  • Provides optimum peak shape and efficiencies
  • Optimized for ACQUITY UPC2 System
  • Achieves faster and more efficient chiral method development and analysis
  • Packed with small particles for added performance
  • Compatible with a full range of mass spectrometers
Catalog No. 50-239-0768 Supplier Waters Corp Supplier No. 186007654
These 2.5 μm columns for chiral separations are designed to reduce method development time while providing optimum peak shape and efficiencies. They are optimized for ACQUITY UPC2 Systems, allowing for faster and more efficient chiral method development and analysis with the added performance of columns packed with small particles and compatibility with a full range of mass spectrometers.
  • Designed to reduce method development time
  • Provides optimum peak shape and efficiencies
  • Optimized for ACQUITY UPC2 System
  • Achieves faster and more efficient chiral method development and analysis
  • Packed with small particles for added performance
  • Compatible with a full range of mass spectrometers