Learn More
Waters Corp Waters Corporation Column, CEL2, 2.5 μm, 3 mm X 50 mm
Supplier:  Waters Corp 186007656
Encompass_Preferred
These 2.5 μm columns are designed for chiral separations, reducing method development time and providing optimum peak shape and efficiencies. Optimized for the ACQUITY UPC2 System, they enable faster and more efficient chiral method development and analysis with small particles compatible with a full range of mass spectrometers.
  • Stationary phase is modified polysaccharide-based.
  • Provides broad-spectrum chiral selectivity.
  • Designed to reduce method development time.
  • Achieves faster and more efficient chiral method development and analysis.
  • Optimized for convergence chromatography.
  • Compatible with a full range of mass spectrometers.
  • Features efficient 2.5 μm particle size chemistries.
  • Attached with an eCord that stores column manufacturing QC data and Certificate of Analysis.
Catalog No. 50-203-8141