Learn More
iQ Biosciences Human Treg Suppression Assay Kit w/ CFSE-Labeled CD8 T Cells
Catalog No. NC2069123
Encompass
Human Treg functional kits with Regulatory T cells and CD8 T cell responders. CD8 T cells are CFSE-labeled for downstream functional readout. Treg cells are validated for regulatory function. Used to test molecules for modulation of Treg function. Carefully cryopreserved to ensure high viability (> 90%) upon thawing. All orders come with an iQ Certificate of Analysis
Catalog No. NC2069123 Supplier iQ Biosciences Supplier No. IQBKA0021C
Add to Cart
Add to Cart
Human Treg functional kits with Regulatory T cells and CD8 T cell responders. CD8 T cells are CFSE-labeled for downstream functional readout. Treg cells are validated for regulatory function. Used to test molecules for modulation of Treg function. Carefully cryopreserved to ensure high viability (> 90%) upon thawing. All orders come with an iQ Certificate of Analysis