Learn More
iQ Biosciences Human Treg Suppression Assay Kit w/ CFSE-Labeled PBMCs
Catalog No. NC2069122
Encompass_Preferred
Human Treg functional kits with Regulatory T cells and PBMC responders. PBMCs are CFSE-labeled for downstream functional readout. Treg cells are validated for regulatory function. Used to test molecules for modulation of Treg function. Carefully cryopreserved to ensure high viability (> 90%) upon thawing. All orders come with an iQ Certificate of Analysis
Catalog No. NC2069122 Supplier iQ Biosciences Supplier No. IQBKA0011C
Human Treg functional kits with Regulatory T cells and PBMC responders. PBMCs are CFSE-labeled for downstream functional readout. Treg cells are validated for regulatory function. Used to test molecules for modulation of Treg function. Carefully cryopreserved to ensure high viability (> 90%) upon thawing. All orders come with an iQ Certificate of Analysis