Learn More
iQ Biosciences Human Treg Suppression Assay Kit w/ CD8 T Cells
Catalog No. NC1820751
Encompass
Human Treg functional kits with Regulatory T cells and CD8 T cell responders. Treg cells are validated for regulatory function. Used to test molecules for modulation of Treg function. Carefully cryopreserved to ensure high viability (> 90%) upon thawing. All orders come with an iQ Certificate of Analysis
Catalog No. NC1820751 Supplier iQ Biosciences Supplier No. IQBKA0021
Human Treg functional kits with Regulatory T cells and CD8 T cell responders. Treg cells are validated for regulatory function. Used to test molecules for modulation of Treg function. Carefully cryopreserved to ensure high viability (> 90%) upon thawing. All orders come with an iQ Certificate of Analysis