Learn More
Aviva Systems Biology Corp TREH Antibody - middle region (ARP65519_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65519_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TREH. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human TREH. Alternate names: MGC129621, TRE, TREAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-512