Learn More
Apexbio Technology LLC Trehalose(Synonyms: Mycose, α,α-Trehalose, alpha-D-glucopyranosyl-alpha-D-glucopyranoside), 100mg, CAS: 99-20-7.
SDP
Catalog No. 5000061267
Encompass_Preferred
Trehalose (CAS 99-20-7) isolated from Saccharomyces cerevisiae is a non-reducing disaccharide composed of two -glucose units connected by an -1 1-glucosidic linkage It enhances cellular resistance to stress factors such as dehydration and cryopreservation however mammalian cells require intracellular presence of trehalose which is limited by low membrane permeability due to its hydrophilic non-ionic nature Derivatization strategies (e g acetylation) to increase membrane traversal may enable the application of trehalose in biopreservation and cytoprotection research
Catalog No. 50-000-61267 Supplier Apexbio Technology LLC Supplier No. B7916100
Trehalose (CAS 99-20-7) isolated from Saccharomyces cerevisiae is a non-reducing disaccharide composed of two -glucose units connected by an -1 1-glucosidic linkage It enhances cellular resistance to stress factors such as dehydration and cryopreservation however mammalian cells require intracellular presence of trehalose which is limited by low membrane permeability due to its hydrophilic non-ionic nature Derivatization strategies (e g acetylation) to increase membrane traversal may enable the application of trehalose in biopreservation and cytoprotection research