Learn More
Cayman Chemical Trehalose 6,6'-Dibehenate, Purity: ≥95%, CAS: 66758-35-8, 10 mg
SDP
Catalog No. 502917677
Encompass_Preferred
A synthetic glycolipid and derivative of TDM; binds to a fusion protein of mouse Mincle and human IgG1 Fc in a cell-free assay at 10 µg/ml; induces NO and G-CSF production in mouse bone marrow macrophages in a Mincle-dependent manner at 5 µg/ml; liposomes containing TDB and DDA as adjuvants and Ac2SGL and PIM2 decrease spleen bacterial load and the number of lung and spleen lesions in a guinea pig model of M. tuberculosis infection
Catalog No. 50-291-7677 Supplier Cayman Chemical Supplier No. 3791110 MG
A synthetic glycolipid and derivative of TDM; binds to a fusion protein of mouse Mincle and human IgG1 Fc in a cell-free assay at 10 µg/ml; induces NO and G-CSF production in mouse bone marrow macrophages in a Mincle-dependent manner at 5 µg/ml; liposomes containing TDB and DDA as adjuvants and Ac2SGL and PIM2 decrease spleen bacterial load and the number of lung and spleen lesions in a guinea pig model of M. tuberculosis infection