Learn More
Medchemexpress LLC Trehalose-6,6'-dibehenate | 66758-35-8 | 99.9% | 987.43 | 10 MG
SDP
Catalog No. 5000435941
Encompass_Preferred
Trehalose-6,6'-dibehenate is an orally active glycolipid that activates Mincle and inflammasome, inducing IL-1β secretion in a caspase activity-dependent manner. It also exerts a Myd88-dependent adjuvant activity and induces Th-1/Th-17 immune responses.
  • Orally active glycolipid
  • Activates Mincle and inflammasome
  • Induces IL-1β secretion in a caspase activity-dependent manner
  • Exerts a Myd88-dependent adjuvant activity
  • Induces Th-1/Th-17 immune responses
Catalog No. 50-004-35941 Supplier Medchemexpress LLC Supplier No. HY11213710MG
Trehalose-6,6'-dibehenate is an orally active glycolipid that activates Mincle and inflammasome, inducing IL-1β secretion in a caspase activity-dependent manner. It also exerts a Myd88-dependent adjuvant activity and induces Th-1/Th-17 immune responses.
  • Orally active glycolipid
  • Activates Mincle and inflammasome
  • Induces IL-1β secretion in a caspase activity-dependent manner
  • Exerts a Myd88-dependent adjuvant activity
  • Induces Th-1/Th-17 immune responses