Learn More
Medchemexpress LLC Trehalose-6,6'-dibehenate | 66758-35-8 | MFCD00057968 | 99.9% | 987.43 g/mol | C56H106O13 | 5 MG
SDP
Catalog No. 5000264257
Encompass_Preferred
Trehalose-6,6'-dibehenate is a bioactive glycolipid research reagent that functions as an immunostimulatory adjuvant. It activates the C-type lectin receptor Mincle and the inflammasome, leading to IL-1β secretion in a caspase-dependent manner and promoting Th1/Th17 immune responses. The material is provided as a high-purity solid with specified storage conditions for long-term stability.
  • Bioactive glycolipid that activates Mincle and inflammasome.
  • Induces IL-1β secretion and promotes Th1/Th17 responses.
  • High purity (>99.9%) white to off-white solid.
  • Molecular weight 987.43 g/mol and formula C56H106O13.
  • Stable when stored as powder at -20°C for extended periods.
  • Supplied in small research quantities appropriate for assay development.
Catalog No. 50-002-64257 Supplier Medchemexpress LLC Supplier No. HY1121375MG
Trehalose-6,6'-dibehenate is a bioactive glycolipid research reagent that functions as an immunostimulatory adjuvant. It activates the C-type lectin receptor Mincle and the inflammasome, leading to IL-1β secretion in a caspase-dependent manner and promoting Th1/Th17 immune responses. The material is provided as a high-purity solid with specified storage conditions for long-term stability.
  • Bioactive glycolipid that activates Mincle and inflammasome.
  • Induces IL-1β secretion and promotes Th1/Th17 responses.
  • High purity (>99.9%) white to off-white solid.
  • Molecular weight 987.43 g/mol and formula C56H106O13.
  • Stable when stored as powder at -20°C for extended periods.
  • Supplied in small research quantities appropriate for assay development.