Learn More
Medchemexpress LLC Trehalose 6,6'-dimycolate | 61512-20-7 | MFCD18643296 | 97.0% | 2053.4 g/mol | C130H250O15 | 5 MG
SDP
Catalog No. 5000283827
Encompass_Preferred
Trehalose 6,6′-dimycolate (cord factor) is a mycobacterial cell-wall glycolipid used as a research reagent to simulate Mycobacterium tuberculosis-induced inflammation and granuloma formation. It is supplied as a powder for laboratory use and has defined storage and handling recommendations to preserve activity.
  • Used to model mycobacterial-induced inflammation and granuloma formation.
  • Cell-wall glycolipid (cord factor) relevant to tuberculosis research.
  • Purity 97.0%.
  • Available in small research pack sizes suitable for in vitro studies.
  • Powder form with recommended cold storage to maintain stability.
  • Applicable for innate immunity and adjuvant mechanism studies.
Catalog No. 50-002-83827 Supplier Medchemexpress LLC Supplier No. HYW1456575MG
Trehalose 6,6′-dimycolate (cord factor) is a mycobacterial cell-wall glycolipid used as a research reagent to simulate Mycobacterium tuberculosis-induced inflammation and granuloma formation. It is supplied as a powder for laboratory use and has defined storage and handling recommendations to preserve activity.
  • Used to model mycobacterial-induced inflammation and granuloma formation.
  • Cell-wall glycolipid (cord factor) relevant to tuberculosis research.
  • Purity 97.0%.
  • Available in small research pack sizes suitable for in vitro studies.
  • Powder form with recommended cold storage to maintain stability.
  • Applicable for innate immunity and adjuvant mechanism studies.