Learn More
Medchemexpress LLC Trehalose 6,6'-dimycolate (cord factor) | 61512-20-7 | MFCD18643296 | 97.0% | 2053.4 g/mol | C130H250O15 | 1 MG
SDP
Catalog No. 5000282922
Encompass_Preferred
Trehalose 6,6′-dimycolate is a mycobacterial cell-wall glycolipid used as a research reagent to model Mycobacterium tuberculosis-induced inflammation and granuloma formation. It modulates innate and adaptive immune responses and is applied in both in vitro and in vivo immunology studies.
  • Mimics mycobacterial cord factor activity.
  • Induces granuloma formation and inflammatory signaling.
  • Modulates macrophage killing and antigen presentation.
  • Suitable for in vitro and in vivo immunology research.
  • Provided as small-quantity, high-purity material (97.0%).
Catalog No. 50-002-82922 Supplier Medchemexpress LLC Supplier No. HYW1456571MG
Trehalose 6,6′-dimycolate is a mycobacterial cell-wall glycolipid used as a research reagent to model Mycobacterium tuberculosis-induced inflammation and granuloma formation. It modulates innate and adaptive immune responses and is applied in both in vitro and in vivo immunology studies.
  • Mimics mycobacterial cord factor activity.
  • Induces granuloma formation and inflammatory signaling.
  • Modulates macrophage killing and antigen presentation.
  • Suitable for in vitro and in vivo immunology research.
  • Provided as small-quantity, high-purity material (97.0%).