Learn More
Medchemexpress LLC α,α-trehalose 6-phosphate (potassium) | 136632-28-5 | MFCD00133871 | 99.0% | C12H21K2O14P | 10MG
SDP
Catalog No. 5000261975
Encompass_Preferred
α,α-Trehalose 6-phosphate potassium is the potassium salt of trehalose 6-phosphate, an endogenous sugar phosphate used as a research reagent for studies of carbohydrate metabolism, enzyme activity, and signaling. It is supplied as a solid, research-grade chemical suitable for laboratory assays and analytical methods.
  • High purity (99.0%).
  • Potassium salt form increases solubility in water.
  • Suitable for enzymatic assays and metabolic pathway studies.
  • Provided as a dry solid for easy storage and weighing.
  • Compatible with aqueous buffers and common biochemical solvents.
  • Store refrigerated to maintain stability.
Catalog No. 50-002-61975 Supplier Medchemexpress LLC Supplier No. HY13136810MG
α,α-Trehalose 6-phosphate potassium is the potassium salt of trehalose 6-phosphate, an endogenous sugar phosphate used as a research reagent for studies of carbohydrate metabolism, enzyme activity, and signaling. It is supplied as a solid, research-grade chemical suitable for laboratory assays and analytical methods.
  • High purity (99.0%).
  • Potassium salt form increases solubility in water.
  • Suitable for enzymatic assays and metabolic pathway studies.
  • Provided as a dry solid for easy storage and weighing.
  • Compatible with aqueous buffers and common biochemical solvents.
  • Store refrigerated to maintain stability.