Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEOGEN Trehalose Assay Kit, Part Number: 700004348, Enzymatic assay for trehalose quantification, Assay method: Spectrophotometer, Range: 4 to 80 μg, Assays: 100 manual, 1000 microplate
SDP

Catalog No. NC2308237
Encompass_Preferred

The NEOGEN Trehalose Assay Kit provides a reliable method for quantifying trehalose in various sample matrices using a spectrophotometric approach. This enzymatic assay is highly specific and adaptable for different laboratory workflows, making it suitable for scientific and industrial applications, including food and pharmaceutical analysis.

  • Simple and rapid assay for trehalose quantification
  • Specific for trehalose via enzymatic hydrolysis
  • Supports multiple assay formats: spectrophotometer, microplate, auto-analyser
  • Detection range: 4 to 80 μg of trehalose per assay
  • Stable for over 2 years under recommended conditions
  • Ambient shipping and 2-8°C storage recommendations
  • Applications in food, beverages, pharmaceuticals, and more

Catalog No. NC2308237 Supplier NEOGEN Supplier No. 700004348
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The NEOGEN Trehalose Assay Kit provides a reliable method for quantifying trehalose in various sample matrices using a spectrophotometric approach. This enzymatic assay is highly specific and adaptable for different laboratory workflows, making it suitable for scientific and industrial applications, including food and pharmaceutical analysis.

  • Simple and rapid assay for trehalose quantification
  • Specific for trehalose via enzymatic hydrolysis
  • Supports multiple assay formats: spectrophotometer, microplate, auto-analyser
  • Detection range: 4 to 80 μg of trehalose per assay
  • Stable for over 2 years under recommended conditions
  • Ambient shipping and 2-8°C storage recommendations
  • Applications in food, beverages, pharmaceuticals, and more

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.