Learn More
Dojindo Molecular Technologies Inc Trehalose C12 detergent (500MG), CAS: 64622-91-9, is used as a stabilizer and protective material for biomaterial compounds
SDP
Catalog No. NC0721618
Encompass
Trehalose is commonly used as a stabilizer and protective material for biomaterial compounds. Because of these characteristics, Trehalose is introduced to the new detergents as a hydrophilic part. Due to the great character of Trehalose, protein degeneration is expected to be prevented with Trehalose detergents. Therefore, Trehalose detergents may keep the proteins E-functions and be utilized in the field of proteomics research. The 5 kinds of Trehalose detergents, with different alkyl chains from 8 to 16, are available. (https://www.dojindo.com/product/trehalose-detergent-t459-t460-t461-t464-t465/)
Catalog No. NC0721618 Supplier Dojindo Molecular Technologies Inc Supplier No. T46110
Trehalose is commonly used as a stabilizer and protective material for biomaterial compounds. Because of these characteristics, Trehalose is introduced to the new detergents as a hydrophilic part. Due to the great character of Trehalose, protein degeneration is expected to be prevented with Trehalose detergents. Therefore, Trehalose detergents may keep the proteins E-functions and be utilized in the field of proteomics research. The 5 kinds of Trehalose detergents, with different alkyl chains from 8 to 16, are available. (https://www.dojindo.com/product/trehalose-detergent-t459-t460-t461-t464-t465/)