Learn More
Cayman Chemical TrelaglIptIn 10mg
SDP
Catalog No. 502967688
Encompass_Preferred
A DPP-4 inhibitor (IC50 = 4 nM)>10,000-fold selective for DPP-4 over isopeptidases; inhibits plasma DPP-4 activity and reduces postprandial plasma insulin levels in Zucker fa/fa rats at 7 mg/kg
Catalog No. 50-296-7688 Supplier Cayman Chemical Supplier No. 1850410 MG
A DPP-4 inhibitor (IC50 = 4 nM)>10,000-fold selective for DPP-4 over isopeptidases; inhibits plasma DPP-4 activity and reduces postprandial plasma insulin levels in Zucker fa/fa rats at 7 mg/kg