Learn More
Selleck Chemical LLC Trelagliptin succinate S5063-25mg
SDP
Catalog No. 503111901
Encompass_Preferred
Trelagliptin succinate (SYR472) is a dipeptidyl peptidase IV (DPP-4) inhibitor which is used as a new long-acting drug for once-weekly treatment of type 2 diabetes mellitus (DM)
Catalog No. 50-311-1901 Supplier Selleck Chemical LLC Supplier No. S506325MG
Trelagliptin succinate (SYR472) is a dipeptidyl peptidase IV (DPP-4) inhibitor which is used as a new long-acting drug for once-weekly treatment of type 2 diabetes mellitus (DM)