Learn More
Apexbio Technology LLC TRELAGLIPTIN-SUCCINATE-50MG
SDP
Catalog No. 5000567983
Encompass_Preferred
Trelagliptin succinate (CAS No 1029877-94-8) is a dipeptidyl peptidase-4 (DPP-4) inhibitor primarily researched in the field of diabetes treatment Originating from efforts to improve glycemic control its mechanism involves prolonging the activity of incretin hormones thereby enhancing insulin secretion and suppressing glucagon release in a glucose-dependent manner In vitro it exhibits activity in the nanomolar to low micromolar range Common applications include studies in cell models animal studies and drug screening focusing on glycemic regulation and metabolic effects Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67983 Supplier Apexbio Technology LLC Supplier No. A388950
Trelagliptin succinate (CAS No 1029877-94-8) is a dipeptidyl peptidase-4 (DPP-4) inhibitor primarily researched in the field of diabetes treatment Originating from efforts to improve glycemic control its mechanism involves prolonging the activity of incretin hormones thereby enhancing insulin secretion and suppressing glucagon release in a glucose-dependent manner In vitro it exhibits activity in the nanomolar to low micromolar range Common applications include studies in cell models animal studies and drug screening focusing on glycemic regulation and metabolic effects Experimental concentrations typically depend on the specific research design