Learn More
Medchemexpress LLC HY-15408A 5mg Medchemexpress, Trelagliptin (succinate) CAS:1029877-94-8 Purity:>98%
SDP
Catalog No. 502081718
Encompass_Preferred
Medchemexpress, HY-15408A 5mg Trelagliptin (succinate) CAS:1029877-94-8 Trelagliptin (SYR-472) succinate is a potent, orally active and highly selective DPP-4 inhibitor with an IC50 of 4 nM. Trelagliptin succinate improves glycemic control in vivo and can be used for the study of type 2 diabetes mellitus (T2DM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1718 Supplier Medchemexpress LLC Supplier No. HY15408A5MG
Medchemexpress, HY-15408A 5mg Trelagliptin (succinate) CAS:1029877-94-8 Trelagliptin (SYR-472) succinate is a potent, orally active and highly selective DPP-4 inhibitor with an IC50 of 4 nM. Trelagliptin succinate improves glycemic control in vivo and can be used for the study of type 2 diabetes mellitus (T2DM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.