Learn More
Sino Biological Anti-TREM-1 Antibody, Rabbit Polyclonal
Catalog No. 5000083807
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TREM-1 (rh TREM-1; Catalog#10511-H08H; NP_061113.1; Met1-Arg200). TREM-1 specific IgG was purified by Human TREM-1 affinity chromatography.
Catalog No. 50-000-83807 Supplier Sino Biological Supplier No. 10511T16100
Produced in rabbits immunized with purified, recombinant Human TREM-1 (rh TREM-1; Catalog#10511-H08H; NP_061113.1; Met1-Arg200). TREM-1 specific IgG was purified by Human TREM-1 affinity chromatography.