Learn More
Enzo Life Sciences TREM-2 monoclonal antibody (2B5) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS7440100
Encompass_Preferred
Clone: 2B5. Isotype: IgG3κ. Immunogen: Recombinant human TREM2 (aa 19-161) purified from E. coli. Source: Purified from ascites. Species reactivity: Human. Applications: ELISA, Flow Cytometry, ICC, IF, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 10% glycerol.
Catalog No. 50-201-3285