Learn More
Sino Biological Human TREM2 Protein (ECD,His Tag) 1mg
Supplier:  Sino Biological 11084H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human TREM2 (NP_061838.1) (Met1-Ser174) was expressed fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6332