Learn More
Cell Signaling Technology Matched Antibody Pair, TREM2, Mouse, Unconjugated, 1 Kit
SDP
Catalog No. 502731861
Encompass
The TREM2 (Full-length) Matched Antibody Pair is designed for use in immunoassay applications, including high-throughput ELISA platforms. This kit provides a validated capture and detection antibody pair optimized for studies requiring specific antibody labeling, such as fluorophores, lanthanides, biotin, or bead-based systems. These antibodies facilitate the detection of TREM2 in mouse samples and are compatible with various immunoassay formats including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Validated capture and detection antibodies that bind to non-overlapping epitopes
  • Supplied in 1X PBS (pH 7.8)
  • BSA and azide free formulation
  • Optimized for mouse species reactivity
  • Suitable for custom labeling technologies
Catalog No. 50-273-1861 Supplier Cell Signaling Technology Supplier No. 71187S
The TREM2 (Full-length) Matched Antibody Pair is designed for use in immunoassay applications, including high-throughput ELISA platforms. This kit provides a validated capture and detection antibody pair optimized for studies requiring specific antibody labeling, such as fluorophores, lanthanides, biotin, or bead-based systems. These antibodies facilitate the detection of TREM2 in mouse samples and are compatible with various immunoassay formats including MSD, Quanterix Simoa, Alpha Technology, and Luminex.
  • Validated capture and detection antibodies that bind to non-overlapping epitopes
  • Supplied in 1X PBS (pH 7.8)
  • BSA and azide free formulation
  • Optimized for mouse species reactivity
  • Suitable for custom labeling technologies