Learn More
Cell Signaling Technology TREM2 Signaling Pathways Antibody Sampler Kit, 9-Antibody Set
SDP
Catalog No. 502731343
Encompass
The TREM2 Signaling Pathways Antibody Sampler Kit provides a collection of rabbit monoclonal antibodies to detect key proteins involved in the TREM2 signaling cascade. This kit is designed for the study of microglial cellular functions, such as phagocytosis, cytokine release, and microgliosis, which are critical in neurodegenerative research including Alzheimer™s disease.
  • Includes nine primary antibodies (20 μL each) and one HRP-linked secondary antibody (100 μL).
  • Validated for multiple applications including Western blotting, immunofluorescence, immunohistochemistry (paraffin), flow cytometry, and immunoprecipitation.
  • Components provided in stable storage buffer.
  • Includes total and phospho-specific antibodies for TREM2, DAP12, Syk, ZAP-70, PLCγ1, and PLCγ2.
Catalog No. 50-273-1343 Supplier Cell Signaling Technology Supplier No. 77533T
The TREM2 Signaling Pathways Antibody Sampler Kit provides a collection of rabbit monoclonal antibodies to detect key proteins involved in the TREM2 signaling cascade. This kit is designed for the study of microglial cellular functions, such as phagocytosis, cytokine release, and microgliosis, which are critical in neurodegenerative research including Alzheimer™s disease.
  • Includes nine primary antibodies (20 μL each) and one HRP-linked secondary antibody (100 μL).
  • Validated for multiple applications including Western blotting, immunofluorescence, immunohistochemistry (paraffin), flow cytometry, and immunoprecipitation.
  • Components provided in stable storage buffer.
  • Includes total and phospho-specific antibodies for TREM2, DAP12, Syk, ZAP-70, PLCγ1, and PLCγ2.