Learn More
Sino Biological TREM-2/TREM2 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11084T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TREM-2/TREM2 (rh TREM-2/TREM2; Catalog#11084-H08H; NP_061838.1; Met1-Ser174). TREM-2/TREM2 specific IgG was purified by Human TREM-2/TREM2 affinity chromatography.
Catalog No. 50-224-7364