Learn More
Aviva Systems Biology Corp TREML2 Antibody - middle region (ARP49878_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49878_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TREML2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TREML2. Alternate names: C6orf76, FLJ13693, MGC149715, MGC149716, TLT2, dJ238O23.1, TLT-2ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-60206