Learn More
Strem, An Ascensus Company CAS# 4097-89-6. 10g. 2,2',2''-Triaminotriethylamine, min. 98% TREN. MFCD00008177
SDP
Catalog No. 502045222
Encompass
CAS# 4097-89-6. 10g. 2,2',2''-Triaminotriethylamine, min. 98% TREN. MFCD00008177. Molecular Weight: 146.24. Molecular Formula: N(CH2CH2NH2)3. Color/form: colorless to pale yellow liq. Strem# 07-2400. http://www.strem.com/catalog/v/07-2400/
Catalog No. 50-204-5222 Supplier Strem, An Ascensus Company Supplier No. 07240010G
CAS# 4097-89-6. 10g. 2,2',2''-Triaminotriethylamine, min. 98% TREN. MFCD00008177. Molecular Weight: 146.24. Molecular Formula: N(CH2CH2NH2)3. Color/form: colorless to pale yellow liq. Strem# 07-2400. http://www.strem.com/catalog/v/07-2400/