Learn More
StatLab™ Treosin Stain
GSA_VASDP
Catalog No. STLSL9316
Change view
Click to view available options
Quantity:
4 x 1 gal.
4 x 16 oz. Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLSL9316 4 x 16 oz. Bottle
STLSL931 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL9316 Supplier StatLab™ Supplier No. SL9316
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Slightly acidified combination of Eosin-Y with Orange-G and Acid Fuchsin.
  • Suitable as replacement for routine eosin
  • Distinct shades of eosin easily obtained from bright red-orange to bright red to red-pink
  • Great for frozen sections
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 4 x 16 oz. Bottle
Product Type Eosin Stain