Learn More
Medchemexpress LLC HY-16503 5mg Medchemexpress, Treosulfan CAS:299-75-2 Purity:>98%
SDP
Catalog No. 502081889
Encompass_Preferred
Medchemexpress, HY-16503 5mg Treosulfan CAS:299-75-2 Treosulfan (NSC 39069) is a bifunctional alkylating agent with activity in ovarian cancer and other solid tumor types. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1889 Supplier Medchemexpress LLC Supplier No. HY165035MG
Medchemexpress, HY-16503 5mg Treosulfan CAS:299-75-2 Treosulfan (NSC 39069) is a bifunctional alkylating agent with activity in ovarian cancer and other solid tumor types. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.